Assay Details
| Gene Symbol: lyz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139180.1 | NP_631919.1 | 2 - 3 | 324 | 127 | |
| GenBank mRNA | AF402599.1 | - | 2 - 3 | 324 | 127 | |
| BC092921.1 | - | 2 - 3 | 341 | 127 | ||
| BC114260.1 | - | 2 - 3 | 324 | 127 | ||
| BC162644.1 | - | 2 - 3 | 321 | 127 | ||
More Information
Set Membership: |
|

lyz
Genomic Map