Assay Details
| Gene Symbol: myo7aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152983.1 | NP_694515.1 | 28 - 29 | 3914 | 97 | |
| XM_021467573.1 | XP_021323248.1 | 29 - 30 | 4065 | 97 | ||
| GenBank mRNA | AJ404001.1 | - | 28 - 29 | 3914 | 97 | |
| AJ404002.1 | - | 27 - 28 | 3794 | 97 | ||
| BC163570.1 | - | 27 - 28 | 3802 | 97 | ||
More Information
Set Membership: |
|

myo7aa
Genomic Map