Assay Details
| Gene Symbol: myo7aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152983.1 | NP_694515.1 | 33 - 34 | 4728 | 81 | |
| XM_021467573.1 | XP_021323248.1 | 34 - 35 | 4879 | 81 | ||
| GenBank mRNA | AJ404001.1 | - | 33 - 34 | 4728 | 81 | |
| AJ404002.1 | - | 32 - 33 | 4608 | 81 | ||
| BC163570.1 | - | 32 - 33 | 4616 | 81 | ||
More Information
Set Membership: |
|

myo7aa
Genomic Map