Assay Details
| Gene Symbol: myo7aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152983.1 | NP_694515.1 | 39 - 40 | 5680 | 70 | |
| XM_021467573.1 | XP_021323248.1 | 40 - 41 | 5831 | 70 | ||
| GenBank mRNA | AJ404001.1 | - | 39 - 40 | 5680 | 70 | |
| AJ404002.1 | - | 38 - 39 | 5560 | 70 | ||
More Information
Set Membership: |
|

myo7aa
Genomic Map