Assay Details
| Gene Symbol: myo7aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152983.1 | NP_694515.1 | 46 - 47 | 6481 | 119 | |
| GenBank mRNA | AJ404001.1 | - | 46 - 47 | 6481 | 119 | |
| BC163570.1 | - | 45 - 46 | 6369 | 119 | ||
More Information
Set Membership: |
|

myo7aa
Genomic Map