Assay Details
| Gene Symbol: ddx19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173258.1 | NP_775365.1 | 11 - 12 | 1531 | 85 | |
| GenBank mRNA | AY099536.1 | - | 11 - 12 | 1531 | 85 | |
| BC044541.1 | - | 11 - 12 | 1612 | 85 | ||
More Information
Set Membership: |
|

ddx19
Genomic Map