Assay Details
| Gene Symbol: nifk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173288.1 | NP_775395.1 | 2 - 3 | 348 | 82 | |
| GenBank mRNA | AF506200.1 | - | 2 - 3 | 348 | 82 | |
| BC071545.1 | - | 2 - 3 | 342 | 82 | ||
More Information
Set Membership: |
|

nifk
Genomic Map