Assay Details
| Gene Symbol: smad3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175083.2 | NP_778258.1 | 8 - 9 | 1154 | 103 | |
| GenBank mRNA | AY134490.1 | - | 8 - 9 | 1154 | 103 | |
| BC081628.1 | - | 8 - 9 | 1292 | 103 | ||
More Information
Set Membership: |
|

smad3b
Genomic Map