Assay Details
| Gene Symbol: epb41b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175084.2 | NP_778259.1 | 10 - 11 | 1264 | 88 | |
| GenBank mRNA | AY124488.1 | - | 10 - 11 | 1264 | 88 | |
| BC162616.1 | - | 10 - 11 | 1192 | 88 | ||
More Information
Set Membership: |
|

epb41b
Genomic Map