Assay Details
| Gene Symbol: selenof | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178294.5 | NP_840079.1 | 3 - 4 | 331 | 89 | |
| GenBank mRNA | AY221263.1 | - | 3 - 4 | 296 | 89 | |
| BC071511.1 | - | 3 - 4 | 308 | 89 | ||
| CN509507.1 | - | 3 - 4 | 336 | 89 | ||
| CT669138.2 | - | 2 - 3 | 647 | 89 | ||
| CT671399.2 | - | 3 - 4 | 286 | 89 | ||
| EH535677.1 | - | 2 - 3 | 658 | 89 | ||
| EH553008.1 | - | 3 - 4 | 370 | 89 | ||
| Gene Symbol: eif4g1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC154122.1 | - | 1583 | 89 | ||
More Information
Set Membership: |
|

selenof
Genomic Map