Assay Details
| Gene Symbol: selenof | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178294.5 | NP_840079.1 | 4 - 5 | 376 | 90 | |
| GenBank mRNA | BC071511.1 | - | 4 - 5 | 353 | 90 | |
| CN509507.1 | - | 4 - 5 | 381 | 90 | ||
| CT669138.2 | - | 1 - 2 | 602 | 90 | ||
| CT671399.2 | - | 4 - 5 | 331 | 90 | ||
| EH535677.1 | - | 1 - 2 | 613 | 90 | ||
| EH553008.1 | - | 4 - 5 | 415 | 90 | ||
More Information
Set Membership: |
|

selenof
Genomic Map