Assay Details
| Gene Symbol: pfkfb4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198816.1 | NP_942111.1 | 2 - 3 | 300 | 63 | |
| XM_005166025.4 | XP_005166082.1 | 2 - 3 | 382 | 63 | ||
| GenBank mRNA | BC053282.1 | - | 2 - 3 | 300 | 63 | |
| BC060931.1 | - | 2 - 3 | 345 | 63 | ||
More Information
Set Membership: |
|

pfkfb4b
Genomic Map