Assay Details
| Gene Symbol: p2rx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198985.1 | NP_945336.1 | 1 - 2 | 141 | 123 | |
| GenBank mRNA | AY292648.1 | - | 1 - 2 | 141 | 123 | |
More Information
Set Membership: |
|

p2rx8
Genomic Map