Assay Details
| Gene Symbol: rnd3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199522.1 | NP_955816.1 | 4 - 5 | 653 | 76 | |
| GenBank mRNA | AY865559.1 | - | 3 - 4 | 326 | 76 | |
| BC059452.1 | - | 4 - 5 | 653 | 76 | ||
More Information
Set Membership: |
|

rnd3a
Genomic Map