Assay Details
| Gene Symbol: rtca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199536.2 | NP_955830.1 | 6 - 7 | 934 | 59 | |
| GenBank mRNA | AY398409.1 | - | 6 - 7 | 934 | 59 | |
| BC046087.1 | - | 6 - 7 | 942 | 59 | ||
More Information
Set Membership: |
|

rtca
Genomic Map