Assay Details
| Gene Symbol: sephs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199701.2 | NP_955995.2 | 8 - 9 | 1222 | 53 | |
| XM_005159006.4 | XP_005159063.1 | 8 - 9 | 1123 | 53 | ||
| GenBank mRNA | BC045302.1 | - | 8 - 9 | 1080 | 53 | |
| BC164487.1 | - | 7 - 8 | 1000 | 53 | ||
More Information
Set Membership: |
|

sephs1
Genomic Map