Assay Details
| Gene Symbol: prelid3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199734.1 | NP_956028.1 | 5 - 6 | 544 | 69 | |
| GenBank mRNA | AY398316.1 | - | 5 - 6 | 523 | 69 | |
| BC045872.1 | - | 5 - 6 | 544 | 69 | ||
| BC067157.1 | - | 5 - 6 | 479 | 69 | ||
More Information
Set Membership: |
|

prelid3b
Genomic Map