Assay Details
| Gene Symbol: slc6a19b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199736.1 | NP_956030.1 | 9 - 10 | 1402 | 62 | |
| GenBank mRNA | BC059804.1 | - | 9 - 10 | 1402 | 62 | |
| BC154319.1 | - | 9 - 10 | 1404 | 62 | ||
More Information
Set Membership: |
|

slc6a19b
Genomic Map