Assay Details
| Gene Symbol: ccnl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199740.1 | NP_956034.1 | 2 - 3 | 394 | 145 | |
| XM_009298635.3 | XP_009296910.1 | 2 - 3 | 376 | 145 | ||
| GenBank mRNA | BC045378.1 | - | 2 - 3 | 394 | 145 | |
| BC165619.1 | - | 2 - 3 | 353 | 145 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_009298635.3 transcripts.
Set Membership: |
|

ccnl1b
Genomic Map