Assay Details
| Gene Symbol: ccnl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199740.1 | NP_956034.1 | 5 - 6 | 683 | 89 | |
| GenBank mRNA | BC045378.1 | - | 5 - 6 | 683 | 89 | |
| BC165619.1 | - | 5 - 6 | 642 | 89 | ||
More Information
Set Membership: |
|

ccnl1b
Genomic Map