Assay Details
| Gene Symbol: ccnl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199740.1 | NP_956034.1 | 6 - 7 | 784 | 84 | |
| XM_009298635.3 | XP_009296910.1 | 7 - 8 | 780 | 84 | ||
| GenBank mRNA | BC045378.1 | - | 6 - 7 | 784 | 84 | |
| BC165619.1 | - | 6 - 7 | 743 | 84 | ||
More Information
Set Membership: |
|

ccnl1b
Genomic Map