Assay Details
| Gene Symbol: mdh1aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309523.2 | NP_001296452.1 | 5 - 6 | 897 | 125 | |
| NM_001316925.1 | NP_001303854.1 | 5 - 6 | 610 | 125 | ||
| NM_199947.3 | NP_956241.2 | 5 - 6 | 610 | 125 | ||
| GenBank mRNA | BC053158.1 | - | 5 - 6 | 533 | 125 | |
| EH453718.1 | - | 5 - 6 | 587 | 125 | ||
| EH579651.1 | - | 5 - 6 | 718 | 125 | ||
More Information
Set Membership: |
|

mdh1aa
Genomic Map