Assay Details
| Gene Symbol: mdh1aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309523.2 | NP_001296452.1 | 8 - 9 | 1281 | 63 | |
| NM_001316925.1 | NP_001303854.1 | 8 - 9 | 994 | 63 | ||
| NM_199947.3 | NP_956241.2 | 8 - 9 | 994 | 63 | ||
| GenBank mRNA | AW281946.1 | - | 1 - 2 | 446 | 63 | |
| AY161294.1 | - | 8 - 9 | 930 | 63 | ||
| AY391437.1 | - | 8 - 9 | 920 | 63 | ||
| BC053158.1 | - | 8 - 9 | 917 | 63 | ||
| EH523710.1 | - | 1 - 2 | 430 | 63 | ||
More Information
Set Membership: |
|

mdh1aa
Genomic Map