Assay Details
| Gene Symbol: txnipa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200087.1 | NP_956381.1 | 4 - 5 | 1057 | 81 | |
| GenBank mRNA | BC049493.1 | - | 4 - 5 | 1057 | 81 | |
| BC067376.1 | - | 4 - 5 | 1037 | 81 | ||
More Information
Set Membership: |
|

txnipa
Genomic Map