Assay Details
| Gene Symbol: garem | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200169.2 | NP_956463.1 | 4 - 5 | 1652 | 75 | |
| GenBank mRNA | BC045411.1 | - | 4 - 5 | 1653 | 75 | |
More Information
Set Membership: |
|

garem
Genomic Map