Assay Details
| Gene Symbol: mif4gda | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200384.1 | NP_956678.1 | 3 - 4 | 308 | 67 | |
| XM_005166179.4 | XP_005166236.3 | 3 - 4 | 620 | 67 | ||
| GenBank mRNA | BC053283.1 | - | 3 - 4 | 308 | 67 | |
More Information
Set Membership: |
|

mif4gda
Genomic Map