Assay Details
| Gene Symbol: lsm14aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200425.2 | NP_956719.2 | 3 - 4 | 532 | 104 | |
| XM_021470461.1 | XP_021326136.1 | 3 - 4 | 610 | 104 | ||
| GenBank mRNA | BC065337.1 | - | 3 - 4 | 532 | 104 | |
More Information
Set Membership: |
|

lsm14aa
Genomic Map