Assay Details
| Gene Symbol: lsm14aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200425.2 | NP_956719.2 | 5 - 6 | 805 | 121 | |
| XM_005163175.2 | XP_005163232.1 | 4 - 5 | 786 | 121 | ||
| GenBank mRNA | BC054659.1 | - | 6 - 7 | 785 | 121 | |
| BC065337.1 | - | 5 - 6 | 805 | 121 | ||
More Information
Set Membership: |
|

lsm14aa
Genomic Map