Assay Details
| Gene Symbol: lsm14aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200425.2 | NP_956719.2 | 7 - 8 | 1149 | 159 | |
| XM_005163175.2 | XP_005163232.1 | 6 - 7 | 1130 | 159 | ||
| XM_021470461.1 | XP_021326136.1 | 8 - 9 | 1308 | 159 | ||
| XM_021470462.1 | XP_021326137.1 | 7 - 8 | 1214 | 159 | ||
| GenBank mRNA | BC065337.1 | - | 7 - 8 | 1149 | 159 | |
More Information
Set Membership: |
|

lsm14aa
Genomic Map