Assay Details
| Gene Symbol: plppr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200456.1 | NP_956750.1 | 5 - 6 | 750 | 68 | |
| GenBank mRNA | BC055144.1 | - | 5 - 6 | 750 | 68 | |
| BC155823.1 | - | 5 - 6 | 846 | 68 | ||
More Information
Set Membership: |
|

plppr2a
Genomic Map