Assay Details
| Gene Symbol: plppr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200456.1 | NP_956750.1 | 6 - 7 | 918 | 76 | |
| GenBank mRNA | BC055144.1 | - | 6 - 7 | 918 | 76 | |
| BC152003.1 | - | 4 - 5 | 964 | 76 | ||
| BC155823.1 | - | 6 - 7 | 1014 | 76 | ||
More Information
Set Membership: |
|

plppr2a
Genomic Map