Assay Details
| Gene Symbol: gopc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200557.1 | NP_956851.1 | 1 - 2 | 416 | 76 | |
| XM_005170172.4 | XP_005170229.1 | 1 - 2 | 495 | 76 | ||
| GenBank mRNA | BC056565.1 | - | 1 - 2 | 416 | 76 | |
More Information
Set Membership: |
|

gopc
Genomic Map