Assay Details
| Gene Symbol: fzd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200561.2 | NP_956855.1 | 2020 | 81 | ||
| GenBank mRNA | BC056573.1 | - | 6 - 7 | 2022 | 81 | |
| BC065361.1 | - | 2020 | 81 | |||
More Information
Set Membership: |
|

fzd6
Genomic Map