Assay Details
| Gene Symbol: abi1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328427.1 | NP_001315356.1 | 9 - 10 | 1384 | 67 | |
| XM_005162489.4 | XP_005162546.1 | 10 - 11 | 1400 | 67 | ||
| XM_005162496.4 | XP_005162553.1 | 8 - 9 | 1205 | 67 | ||
| XM_005162498.4 | XP_005162555.1 | 8 - 9 | 1117 | 67 | ||
| XM_005162499.4 | XP_005162556.1 | 7 - 8 | 1102 | 67 | ||
| XM_009297227.3 | XP_009295502.1 | 11 - 12 | 1503 | 67 | ||
| XM_009297228.3 | XP_009295503.1 | 10 - 11 | 1487 | 67 | ||
| XM_009297229.3 | XP_009295504.1 | 9 - 10 | 1220 | 67 | ||
| GenBank mRNA | BC059524.1 | - | 9 - 10 | 1121 | 67 | |
More Information
Set Membership: |
|

abi1a
Genomic Map