Assay Details
| Gene Symbol: fscn2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200770.2 | NP_957064.2 | 1 | 595 | 80 | |
| GenBank mRNA | BC059601.1 | - | 1 | 848 | 80 | |
| EU580143.1 | - | 1 | 595 | 80 | ||
More Information
Set Membership: |
|

fscn2b
Genomic Map