Assay Details
| Gene Symbol: snrkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200833.1 | NP_957127.1 | 3 - 4 | 1082 | 67 | |
| XM_005174310.4 | XP_005174367.1 | 3 - 4 | 836 | 67 | ||
| GenBank mRNA | BC060922.1 | - | 3 - 4 | 1082 | 67 | |
More Information
Set Membership: |
|

snrkb
Genomic Map