Assay Details
| Gene Symbol: prdm9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200902.1 | NP_957196.1 | 3 - 4 | 517 | 94 | |
| GenBank mRNA | BC064665.1 | - | 3 - 4 | 517 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

prdm9| Gene Symbol: prdm9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200902.1 | NP_957196.1 | 3 - 4 | 517 | 94 | |
| GenBank mRNA | BC064665.1 | - | 3 - 4 | 517 | 94 | |
Set Membership: |
|