Assay Details
| Gene Symbol: prdm9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200902.1 | NP_957196.1 | 4 - 5 | 572 | 139 | |
| GenBank mRNA | BC064665.1 | - | 4 - 5 | 572 | 139 | |
More Information
Set Membership: |
|

prdm9
Genomic Map