Assay Details
| Gene Symbol: ttc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002122.2 | NP_001002122.2 | 5 - 6 | 794 | 99 | |
| GenBank mRNA | BC049428.1 | - | 5 - 6 | 766 | 99 | |
| BC071463.1 | - | 5 - 6 | 743 | 99 | ||
| EH459652.1 | - | 5 - 6 | 619 | 99 | ||
| EH504663.1 | - | 5 - 6 | 807 | 99 | ||
More Information
Set Membership: |
|

ttc4
Genomic Map