Assay Details
| Gene Symbol: cds2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201186.1 | NP_957480.1 | 10 - 11 | 1089 | 65 | |
| GenBank mRNA | BC048045.1 | - | 10 - 11 | 1033 | 65 | |
| BC055505.1 | - | 10 - 11 | 1089 | 65 | ||
| BC165336.1 | - | 10 - 11 | 1013 | 65 | ||
More Information
Set Membership: |
|

cds2
Genomic Map