Assay Details
| Gene Symbol: nt5c2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328500.1 | NP_001315429.1 | 5 - 6 | 457 | 71 | |
| XM_005156779.4 | XP_005156836.1 | 6 - 7 | 503 | 71 | ||
| XM_021480521.1 | XP_021336196.1 | 4 - 5 | 457 | 71 | ||
| GenBank mRNA | BC049497.1 | - | 5 - 6 | 71 | ||
| CK028809.1 | - | 2 - 3 | 187 | 71 | ||
| EB931124.1 | - | 4 - 5 | 389 | 71 | ||
More Information
Set Membership: |
|

nt5c2a
Genomic Map