Assay Details
| Gene Symbol: nt5c2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328500.1 | NP_001315429.1 | 8 - 9 | 705 | 129 | |
| XM_005156779.4 | XP_005156836.1 | 9 - 10 | 751 | 129 | ||
| XM_021480521.1 | XP_021336196.1 | 7 - 8 | 705 | 129 | ||
| GenBank mRNA | BC049497.1 | - | 8 - 9 | 129 | ||
| CK028809.1 | - | 5 - 6 | 435 | 129 | ||
More Information
Set Membership: |
|

nt5c2a
Genomic Map