Assay Details
| Gene Symbol: prim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201448.1 | NP_958856.1 | 5 - 6 | 703 | 73 | |
| GenBank mRNA | BC047856.1 | - | 5 - 6 | 703 | 73 | |
| BC153661.1 | - | 5 - 6 | 625 | 73 | ||
More Information
Set Membership: |
|

prim1
Genomic Map