Assay Details
| Gene Symbol: etf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201460.2 | NP_958868.1 | 2 - 3 | 278 | 96 | |
| XM_021479101.1 | XP_021334776.1 | 2 - 3 | 293 | 96 | ||
| GenBank mRNA | AY398343.1 | - | 2 - 3 | 278 | 96 | |
| BC044515.1 | - | 2 - 3 | 272 | 96 | ||
| BC066583.1 | - | 2 - 3 | 196 | 96 | ||
More Information
Set Membership: |
|

etf1b
Genomic Map