Assay Details
| Gene Symbol: sh3gl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201497.1 | NP_958905.1 | 1 - 2 | 273 | 65 | |
| XM_005166793.4 | XP_005166850.1 | 1 - 2 | 335 | 65 | ||
| XM_009303889.3 | XP_009302164.1 | 1 - 2 | 335 | 65 | ||
| XM_021478204.1 | XP_021333879.1 | 1 - 2 | 389 | 65 | ||
| GenBank mRNA | BC045942.1 | - | 1 - 2 | 273 | 65 | |
| BC164167.1 | - | 1 - 2 | 73 | 65 | ||
More Information
Set Membership: |
|

sh3gl1b
Genomic Map