Assay Details
| Gene Symbol: sh3gl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201497.1 | NP_958905.1 | 2 - 3 | 346 | 111 | |
| XM_005166793.4 | XP_005166850.1 | 2 - 3 | 408 | 111 | ||
| XM_009303889.3 | XP_009302164.1 | 2 - 3 | 408 | 111 | ||
| XM_021478204.1 | XP_021333879.1 | 2 - 3 | 462 | 111 | ||
| GenBank mRNA | BC045942.1 | - | 2 - 3 | 346 | 111 | |
| BC164167.1 | - | 2 - 3 | 146 | 111 | ||
More Information
Set Membership: |
|

sh3gl1b
Genomic Map