Assay Details
| Gene Symbol: sh3gl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201497.1 | NP_958905.1 | 7 - 8 | 956 | 73 | |
| XM_005166793.4 | XP_005166850.1 | 7 - 8 | 1018 | 73 | ||
| XM_009303889.3 | XP_009302164.1 | 7 - 8 | 1018 | 73 | ||
| XM_021478204.1 | XP_021333879.1 | 7 - 8 | 1072 | 73 | ||
| GenBank mRNA | BC045942.1 | - | 7 - 8 | 956 | 73 | |
| BC164167.1 | - | 7 - 8 | 756 | 73 | ||
More Information
Set Membership: |
|

sh3gl1b
Genomic Map