Assay Details
| Gene Symbol: sh3gl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201497.1 | NP_958905.1 | 8 - 9 | 1096 | 80 | |
| GenBank mRNA | BC045942.1 | - | 8 - 9 | 1096 | 80 | |
| BC164167.1 | - | 8 - 9 | 896 | 80 | ||
More Information
Set Membership: |
|

sh3gl1b
Genomic Map