Assay Details
| Gene Symbol: tmem30ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205560.2 | NP_991123.2 | 6 - 7 | 1097 | 65 | |
| GenBank mRNA | BC044384.1 | - | 6 - 7 | 996 | 65 | |
| BC045515.1 | - | 6 - 7 | 996 | 65 | ||
| BC065436.1 | - | 6 - 7 | 1097 | 65 | ||
| BC164770.1 | - | 6 - 7 | 948 | 65 | ||
More Information
Set Membership: |
|

tmem30ab
Genomic Map