Assay Details
| Gene Symbol: ifnphi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207640.1 | NP_997523.1 | 1 - 2 | 167 | 84 | |
| GenBank mRNA | AB093588.1 | - | 2 - 3 | 175 | 84 | |
| AB128865.1 | - | 1 - 2 | 86 | 84 | ||
| AJ544821.1 | - | 1 - 2 | 137 | 84 | ||
| AJ544822.1 | - | 1 - 2 | 137 | 84 | ||
| AY135716.1 | - | 1 - 2 | 137 | 84 | ||
| BC162493.1 | - | 1 - 2 | 150 | 84 | ||
| EF014960.1 | - | 2 - 3 | 121 | 84 | ||
More Information
Set Membership: |
|

ifnphi1
Genomic Map